Anti-AIF1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX89367
89366-984EA 979.96 CAD
89366-984
Anti-AIF1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to AIF1 / IBA1 (isoform 3)

Peptide ELISA: antibody detection limit dilution 1:32000.

Western blot: Approx 17kDa band observed in Human Brain (Frontal Cortex) and Peripheral Blood Lymphocytes lysates (calculated MW of 16.7kDa according to NP_001614.3). Recommended concentration: 0.5-1.5µg/ml.

IHC-P: In paraffin embedded Mouse Brain shows staining of astrocytes. Recommended concentration, 2-4µg/ml.

Type: Primary
Antigen: AIF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR