Anti-AIRE Goat Polyclonal Antibody

Supplier: GeneTex

GTX13573
89359-756EA 742.11 CAD
89359-756
Anti-AIRE Goat Polyclonal Antibody
Antibodies
The function of the protein is not well defined, however it contains zinc finger motifs suggestive of a transcription factor. The protein (isoform 1) is localized to both the nucleus and cytoplasm. Three splice variant mRNAs products have been described [1]. The longer AIRE1 mRNA appears to be more abundant and includes exons 1 through 14. Splice variant AIRE2 includes a portion of the non-coding region of exon 1, an alternatively spliced longer exon 8, plus exons 9 through 14. Variant AIRE3 includes the same exon 1-8-9 sequences as found in AIRE2 but utilizes additional alternative splicing in exon 10 that shifts the reading frame such that a stop codon in exon 12 is utilized. The resulting protein products of these splice variants differ significantly. Defects in this gene cause the autosomal-recessive systemic autoimmune disease termed autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED).

Appr. 60kDa in transfected HEK293 lysates (calculated MW of 57.7kDa according to NP_000374.1). Recommended concentration: 0.01-0.03µg/ml. In paraffin embedded Human Thymic medula shows staining of select nuclei. Recommended concentration, 2.5µg/ml. This antibody is expected to recognize isoforms 1 and 2 (NP_000374.1 and NP_000649.1 respecively).

Type: Primary
Antigen: AIRE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR