Anti-AKAP10 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX106430
89354-686EA 623.96 CAD
89354-686
Anti-AKAP10 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to AKAP10 (A kinase (PRKA) anchor protein 10)

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: AKAP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR