Anti-AKT1 Rabbit Monoclonal Antibody

GTX61033
89363-772EA 556.26 CAD
89363-772
Anti-AKT1 Rabbit Monoclonal Antibody
Akt1/PKBa is a Ser/Thr protein kinase whose kinase activity plays a key role in various cellular functions, including apoptosis, glycogen synthesis, and cell growth . Akt1/PKBa is activated by various growth and survival factors . Phosphorylation on Thr308, Ser473 and Tyr474 is required for full activity. Akt1/PKBa inhibits apoptosis by phosphorylating and inactivating several targets, including caspase-9 . Akt1/PKBa regulates glycogen synthesis by phosphorylating and inactivating glycogen synthase kinase-3a and b . Three mammalian Akt1/PKBa isoforms have been identified.

Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR