Anti-ADH Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX40689
89363-270EA 924.06 CAD
89363-270
Anti-ADH Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Alcohol Dehydrogenase (Biotin)

Dot: Use at an assay dependent dilution. ELISA: 1/1000 - 1/5000. IM: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 40 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ADH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR