Anti-AGP1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX40579
89363-248EA 484.38 CAD
89363-248
Anti-AGP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Alpha 1 Acid Glycoprotein (HRP)

Dot: Use at an assay dependent dilution. Electron Microscopy: Use at an assay dependent dilution. ELISA: 1:20,000. WB: 1:1,000 - 1:5,000. IHC: 1:500 - 1:2,500. Predicted molecular weight: 24 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: AGP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR