Anti-AACT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29374
89352-608EA 609.77 CAD
89352-608
Anti-AACT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to alpha 1 Antichymotrypsin

IHC: This antibody may be diluted to a titer of 1:25-1:50 in an ABC method. Staining Protocol: We suggest an incubation period of 30-60 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation conditions and antibody dilutions should be determined by the user.

Type: Primary
Antigen: AACT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR