Anti-AACT Mouse Monoclonal Antibody [clone: 84]

GTX40821
89363-324EA 502.37 CAD
89363-324
Anti-AACT Mouse Monoclonal Antibody [clone: 84]
The protein is a plasma protease inhibitor and member of the serine protease inhibitor class. Polymorphisms in this protein appear to be tissue specific and influence protease targeting. Variation in this protein's sequence have been implicated in Alzheimer's disease, and deficiency of this protein has been associated with liver disease. Mutations have been identified in patients with Parkinson disease and chronic obstructive pulmonary disease.

Type: Primary
Antigen: AACT
Clonality: Monoclonal
Clone: 84
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR