Anti-CTNNA1 Rabbit Monoclonal Antibody

GTX61621
89364-080EA 556.26 CAD
89364-080
Anti-CTNNA1 Rabbit Monoclonal Antibody
Cadherin and catenin compose cell adhesion complex and are indispensable for tight cell-cell adhesion. Dysfunction of this adhesion complex causes dissociation of cancer cells from primary tumor nodules, thus possibly contributing to cancer invasion and metastasis . At least three proteins (alpha, beta, and gamma catenin) comprise the cytoplasmic domain of the cadherin cell-cell adhesion complex. Data, with the reported structure of other catenin genes, suggest that vinculin and alpha-catenin generate a superfamily of proteins mediating membrane-cytoskeletal associations . Presenilin-1 (PS1) overexpression in human kidney cells enhances cell-cell adhesion and data show that PS1 incorporates into the cadherin/catenin adhesion system and regulates cell-cell adhesion. PS1 concentrates at intercellular contacts in epithelial tissue; in brain, it forms complexes with both E- and N-cadherin and concentrates at synaptic adhesions .

Type: Primary
Antigen: CTNNA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR