Anti-CTNNA1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX111095
89356-568EA 623.96 CAD
89356-568
Anti-CTNNA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to alpha 1 Catenin (catenin alpha-1)

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CTNNA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR