Anti-MAN1A1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX40044
89363-074EA 968.07 CAD
89363-074
Anti-MAN1A1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to alpha 1,2 Mannosidase IA

Type: Primary
Antigen: MAN1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR