Anti-alpha-2-HS-Glycoprotein Sheep Polyclonal Antibody

Supplier: GeneTex
GTX41201
89352-860EA 562.4 CAD
89352-860
Anti-alpha-2-HS-Glycoprotein Sheep Polyclonal Antibody
Antibodies
Sheep polyclonal to Alpha 2 HS Glycoprotein

RID: 5 microl antiserum/cm2 in gel vs 5 microl serum, NT - 1:10. Ie: 100 microl antiserum vs 5 microl serum. DID: 10 microl antiserum vs 2 microl serum. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR