Anti-A2M Chicken Polyclonal Antibody

Supplier: GeneTex

GTX16421
89367-464EA 895.72 CAD
89367-464
Anti-A2M Chicken Polyclonal Antibody
Antibodies
Alpha 2 Macroglobulin is a serum protein which functions as a broad range irreversible proteinase inhibitor that forms a ""trap"" around most proteases. Alpha 2 Macroglobulin is so large a molecule that is tends to remain intravascular. Localized on the surface of peripheral blood lymphocytes. A2M is always present in high levels in circulation.

For ELISA: Use at an assay dependent dilution. For WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR