Anti-ACT1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11007
89356-172EA 996.89 CAD
89356-172
Anti-ACT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to alpha Actinin

IF: Use at a dilution of 1/500. WB: Use at an assay dependent dilution. Predicted molecular weight: 107 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ACT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR