Anti-AP2A2 Mouse Monoclonal Antibody [clone: 100/2]

GTX11326
89357-412EA 514.41 CAD
89357-412
Anti-AP2A2 Mouse Monoclonal Antibody [clone: 100/2]
Mouse monoclonal [100 / 2] to alpha Adaptin

WB: Use at a dilution of 1/200. Reacts in bovine and rat brain preparations, with the 105 and 110 ?-subunits of adaptor complex AP2. The antibody reacts with polypeptides of approximately 100 kDa in bovine liver, human heart fibroblasts, and Madin-Darby bovine kidney cultured cells (MDBK), but not with any components in the 110-115 kDa range from these sources or from rat pheochromocytoma cultured cells (PC12), neuroblastoma or astrocytes. The antibody stains the intact alpha adaptor, as well as the 37 and 40 kDa fragments, but not the 63-66 kDa group of alpha fragments obtained by trypsin cleavage of the alpha subunits molecules. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: AP2A2
Clonality: Monoclonal
Clone: 100/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR