Anti-CTNNA2 Rabbit Monoclonal Antibody

GTX61639
89364-096EA 556.26 CAD
89364-096
Anti-CTNNA2 Rabbit Monoclonal Antibody
It has been shown that alpha N-catenin, a linker between cadherin adhesion receptors and the actin cytoskeleton, is essential for stabilizing dendritic spines in rodent hippocampal neurons in culture. Results suggest that alpha N-catenin is a key regulator for the stability of synaptic contacts . We show here that antibodies against alpha catenin also immunoprecipitate complexes that contain human N-cadherin, mouse P-cadherin, chicken A-CAM (adherens junction-specific CAM; also called N-cadherin) or Xenopus U-cadherin, demonstrating that alpha catenin is complexed with other cadherins .

Type: Primary
Antigen: CTNNA2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR