Anti-PRKAA1 Rabbit Monoclonal Antibody [clone: Y365]

GTX61229
89363-886EA 520.01 CAD
89363-886
Anti-PRKAA1 Rabbit Monoclonal Antibody [clone: Y365]
Rabbit monoclonal antibody [Y365] to AMPK alpha 1

Type: Primary
Antigen: PRKAA1
Clonality: Monoclonal
Clone: Y365
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR