Anti-Amyloid A Mouse Monoclonal Antibody [clone: mcl]

Supplier: GeneTex

GTX27505
89362-594EA 565.61 CAD
89362-594
Anti-Amyloid A Mouse Monoclonal Antibody [clone: mcl]
Antibodies
Type: Primary
Antigen: Amyloid A
Clonality: Monoclonal
Clone: [mcl]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR