Anti-AP2A Goat Polyclonal Antibody

Supplier: Genetex

GTX23707
89361-376EA 640.15 CAD
89361-376
Anti-AP2A Goat Polyclonal Antibody
Antibodies
The AP2 alpha proteins are found in the AP2 complex in clathrin-coated vesicles. The AP2 complex is a heterotetramer consisting of two large adaptins (alpha or beta), a medium adaptin (mu), and a small adaptin (sigma). The complex is part of the protein coat on the cytoplasmic face of coated vesicles, which link clathrin to receptors in vesicles.

Type: Primary
Antigen: AP2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR