Anti-APC2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX22976
89351-840EA 565.28 CAD
89351-840
Anti-APC2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Apc2

Western Blotting: use at a concentration of 5?g / ml for 2hrs at RT. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of ~ 92 kDa, which corresponds to the predicted molecular weight of APC 2.

Type: Primary
Antigen: APC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR