Anti-ATG5 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX102361
89349-556EA 623.96 CAD
89349-556
Anti-ATG5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ATG5 (ATG5 autophagy related 5 homolog (S. cerevisiae))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR