Anti-APOA1 Sheep Polyclonal Antibody

GTX74145
89365-392EA 414.08 CAD
89365-392
Anti-APOA1 Sheep Polyclonal Antibody
Antisera to human apolipoprotein A1 were raised by repeated immunization of sheep with highly purified antigen.An immunogoublin-rich fraction was prepared from whole serum by ammonium sulphate precipitation followed by anion exchange chromotography.

Type: Primary
Antigen: APOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR