Anti-APOB Mouse Monoclonal Antibody [clone: 166]

Supplier: GeneTex

GTX40624
89363-264EA 565.92 CAD
89363-264
Anti-APOB Mouse Monoclonal Antibody [clone: 166]
Antibodies
Mouse monoclonal antibody [1606] to Apolipoprotein B

EIA: Use at an assay dependent dilution. RIA: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: APOB
Clonality: Monoclonal
Clone: 166
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR