Anti-APP Mouse Monoclonal Antibody [clone: 4C3]

Supplier: GeneTex

GTX84873
89366-804EA 747.28 CAD
89366-804
Anti-APP Mouse Monoclonal Antibody [clone: 4C3]
Antibodies
Mouse monoclonal antibody [4C3] to APP

Recommended Starting Dilutions:WB 1:2000 IHC 1:50 ICC/IF 1:100 Flow 1:100Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: APP
Clonality: Monoclonal
Clone: 4C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR