Anti-AQP5 Rabbit Monoclonal Antibody

Supplier: Genetex
GTX62313
89364-392EA 585.93 CAD
89364-392
Anti-AQP5 Rabbit Monoclonal Antibody
Aquaporin 5 is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 5 plays a role in the generation of saliva, tears and pulmonary secretions . It is expressed in epithelia of lung, cornea, and various secretory glands; sites where extracellular osmolality is known to fluctuate . Expression of Aquaporin 5 may have pathophysiological implications attributed to a dysfunctional water regulation by Aquaporin 5such as endolymphatic hydrops (i.e. in Meniere's disease) or sensorineural hearing loss (i.e. in Sjogren's syndrome) . Up-regulation of Aquaporin 5 may be involved in differentiation of human gastric cancer cells .

Type: Primary
Antigen: AQP5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR