Anti-ARA9 Mouse Monoclonal Antibody [clone: 35-2]

Supplier: GeneTex

GTX20468
89367-522EA 742.11 CAD
89367-522
Anti-ARA9 Mouse Monoclonal Antibody [clone: 35-2]
Antibodies
Ah receptor-associated protein (ARA9) is also known as AIP and XAP2. This protein displays structural similarity to the glucocorticoid receptor-associated immunophilin FKBP52. Although they share significant homology, they have distinct cellular roles and biochemical properties. ARA9 is known to affect the potency and efficacy of AHR agonists in the yeast S. cerevisiae as well as having functional consequences on AHR signal transduction.

For WB: Use at a dilution of 1:1,000. This antibody may be used for Western blotting where a band is seen at 37 kDa, representing ARA9. It is specific for the FKBP domain. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: ARA9
Clonality: Monoclonal
Clone: 35-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR