Anti-ARG1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX40551
89363-220EA 484.38 CAD
89363-220
Anti-ARG1 Rabbit Polyclonal Antibody
Antibodies
Peroxidase-conjugated Rabbit Polyclonal antibody to Arginase

Recommended Starting Dilutions:WB: Use at 1:1,000 - 1:5,000ELISA: Use at 1:10,000 - 1:40,000

Type: Primary
Antigen: ARG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR