Anti-SEPT4 Mouse Monoclonal Antibody [clone: ARTS51]

Supplier: GeneTex

GTX10440
89350-320EA 693.24 CAD
89350-320
Anti-SEPT4 Mouse Monoclonal Antibody [clone: ARTS51]
Antibodies
Mouse monoclonal [ARTS51] to ARTS

ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. WB: Use at a dilution of 1/0.5 - 1/1.0. Detects a band of approximately 34 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: SEPT4
Clonality: Monoclonal
Clone: ARTS51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR