Anti-ARVP Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX74194
89365-410EA 742.11 CAD
89365-410
Anti-ARVP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Arginine Vasopressin

Recommended Starting Dilutions: Immunohistology - Frozen: 1/500 - 1/5000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ARVP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR