Anti-ATF2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX59562
89363-718EA 925.91 CAD
89363-718
Anti-ATF2 Rabbit Polyclonal Antibody
Antibodies
The transcription factor ATF-2 (also called CRE-BP1) is a member of the ATF/CREB family of leucine zipper proteins. ATF-2 forms homodimers and heterodimers with c-Jun to initiate CRE-dependent transcription. Phosphorylation of ATF-2 at Thr69 and Thr71 by stress-activated kinases is necessary for transcriptional activation. Myc also induces phosphorylation of ATF-2 at Thr69 and Thr71 to prolong the half-life of ATF-2. ATF-2 also functions as a histone acetyltransferase (HAT) by specifically acetylating histones H2B and H4 in vitro.

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR