Anti-ATG12 Rabbit Monoclonal Antibody

GTX62939
89364-612EA 556.26 CAD
89364-612
Anti-ATG12 Rabbit Monoclonal Antibody
Autophagy is a process of bulk protein degradation in which cytoplasmic components, including organelles, are enclosed in double-membrane structures called autophagosomes and delivered to lysosomes or vacuoles for degradation. Atg12 is the human homolog of a yeast protein involved in autophagy. It is essential for Atg12 conjugation, and the ATG12-ATG5 conjugate is necessary to form autophagic vesicles. The E3-like activity of the Atg12-Atg5 conjugate potentiates the formation of Atg8-PE; making the autophagic pathway analogous to ubiquitin-like pathways where an E3 conjugation ligase is required.

Type: Primary
Antigen: ATG12
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR