Anti-ATG5 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX48583
89363-668EA 499.2 CAD
89363-668
Anti-ATG5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATG5

immunohistochemistry 2.5ug/ml, immunofluorescence , Western Blot 1:500

Type: Primary
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR