Anti-ATG7 Rabbit Monoclonal Antibody

GTX61647
89364-100EA 556.26 CAD
89364-100
Anti-ATG7 Rabbit Monoclonal Antibody
Autophagy is a process of bulk degradation of cytoplasmic components by the lysosome/vacuole and has a significant relationship to several neurodegenerative disorders and myopathies in mammals . Apg7 (Atg7) is a unique E1 enzyme which is essential for both the Apg12p- and Apg8p-modification systems, and plays indispensable roles in autophagy . A new molecular pathway has been defined in which activation of the receptor-interacting protein (a serine-threonine kinase) and Jun amino-terminal kinase induced cell death with the morphology of autophagy. Autophagic death required Atg7 and beclin 1 and was induced by caspase-8 inhibition .

Type: Primary
Antigen: ATG7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR