Anti-ATG7 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX113613
89357-646EA 623.96 CAD
89357-646
Anti-ATG7 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ATG7 (ATG7 autophagy related 7 homolog (S. cerevisiae))

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ATG7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR