Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]

Supplier: GeneTex
GTX30636
89367-684EA 863.11 CAD
89367-684
Anti-ATM Mouse Monoclonal Antibody [clone: 10H11.E12]
Antibodies
Mouse monoclonal antibody to ATM Kinase (phospho Ser1981)

Recommended Starting Dilutions: For WB: Use at - 1/1,000 (ECL) For ICC/IF: Use at 1/1,000 For IP: Use at an assay-dependent dilution IHC-P : 1:200 Optimal dilutions should be determined experimentally by the researcher. Western blotting (a single~370 kDa band should be seen).

Type: Primary
Antigen: ATM
Clonality: Monoclonal
Clone: 10H11.E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR