Anti-ATP6V0A1 Rabbit Polyclonal Antibody

GTX46426
89363-608EA 643.76 CAD
89363-608
Anti-ATP6V0A1 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to ATP6V0A1 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: ATP6V0A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR