Anti-ATP6V1E1 Chicken Polyclonal Antibody

GTX82325
89368-070EA 628.5 CAD
89368-070
Anti-ATP6V1E1 Chicken Polyclonal Antibody
Chicken polyclonal to Vacuolar ATP synthase subunit E

WB (only tested on fusion protein): Use at a dilution of 1:2000. Colorimetric method for signal development.

Type: Primary
Antigen: ATP6V1E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR