Anti-ATP7B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX30640
89352-716EA 888.67 CAD
89352-716
Anti-ATP7B Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ATP7b

By Western blot, this antibody recognizes a band at 165 kDa, representing ATP7b protein. This antibody also recognizes a band at ~220 kDa. This antibody may work on endogeneous protein in immunohistochemistry but it has only been tested on transfected ovarian carcinoma cells at a 1:500 dilution. However, GTX30640 may demonstrate less background for this assay.Suggested working dilutions: *Western Blot - 1:1,000 Immunoprecipitation - NDImmunohistochemistry - (see statement above)*The investigator should determine the optimal working dilution for a specific application.

Type: Primary
Antigen: ATP7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR