Anti-AURKB Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX85607
89366-888EA 753.5 CAD
89366-888
Anti-AURKB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to AURKB (phospho Thr232)

WB (1:250 - 1:2000), ELISA (1:10000 - 1:50000), IF, IHC-P (2.5 µg/ml)

Type: Primary
Antigen: AURKB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR