Anti-AUR1 Rabbit Monoclonal Antibody

GTX61403
89363-970EA 556.26 CAD
89363-970
Anti-AUR1 Rabbit Monoclonal Antibody
Aurora-B is a member of a novel family of serine/threonine kinases that have been identified as key regulators of the mitotic cell division process. Aurora-B is known to be involved in the regulation of centrosome function, bipolar spindle assembly and chromosome segregation. Aurora kinases are localized at the centrosomes of interphase cells, at the poles of the bipolar spindle and in the midbody of the mitotic apparatus. Substrates identified for the Aurora-B kinases include a kinesin-like motor protein, spindle apparatus proteins, histone H3 protein, kinetochore protein and the tumor suppressor protein p53 . Overexpression of aurora B produced multinuclearity and induced aggressive metastasis, suggesting that overexpressed aurora B has multiple functions in cancer development. Identification of Aurora kinases as RasGAP Src homology 3 domain binding protein, also implicates these kinases as potential effectors in the Ras pathway relevant to oncogenesis .

Type: Primary
Antigen: AUR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR