Anti-BACE Rabbit Monoclonal Antibody

GTX62419
89364-430EA 556.26 CAD
89364-430
Anti-BACE Rabbit Monoclonal Antibody
Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer's disease. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein (APP) by two proteases, one of which is BACE1. BACE1 is a member of the peptidase A1 protein family, a type I integral membrane glycoprotein and aspartic protease that is found mainly in the Golgi .

Type: Primary
Antigen: BACE
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR