Anti-ATPA Mouse Monoclonal Antibody

Supplier: GeneTex

GTX28240
89352-346EA 565.64 CAD
89352-346
Anti-ATPA Mouse Monoclonal Antibody
Antibodies
Protective antigen (PA) is one of the three proteins composing the anthrax toxin, the agent which infects many mammalian species and that may cause death. PA binds to a receptor (ATR) in sensitive eukaryotic cells, thereby facilitating the translocation of the enzymatic toxin components, edema factor and lethal factor, across the target cell membrane. PA associated with LF causes death when injected, PA associated with EF produces edema. PA induces immunity to infection with anthrax.

Type: Primary
Antigen: ATPA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR