Anti-BAD Rabbit Polyclonal Antibody

GTX74262
89367-768EA 535.8 CAD
89367-768
Anti-BAD Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Bad

Recommended Starting Dilutions: Western Blotting: Max Dilution 1/1000 Immunohistology - Paraffin: 1/200 - 1/1000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: BAD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR