Anti-BARX2 Mouse Monoclonal Antibody [clone: BAR X2 8A7/1]

Supplier: GeneTex
GTX40128
89363-102EA 673.29 CAD
89363-102
Anti-BARX2 Mouse Monoclonal Antibody [clone: BAR X2 8A7/1]
Antibodies
Mouse monoclonal [BAR X2 8A7/1] to BarX2

ELISA: Use at an assay dependent dilution. IF: 1/500. IHC-P: 1/2. Note: sections need microwave pre-treatment. WB: Use at an assay dependent dilution. Predicted molecular weight: 31 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: BARX2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR