Anti-BDNF Chicken Polyclonal Antibody

Supplier: Genetex

GTX17884
89360-334EA 624.35 CAD
89360-334
Anti-BDNF Chicken Polyclonal Antibody
Antibodies
Brain derived neurotrophic factor (BDNF) is a member of the neurotrophin family of growth factors that includes NGF, NT3, and NT4. All neurotrophins have six conserved cysteine residues and share a 55% sequence identity at the amino acid level. BDNF has been shown to enhance the survival and differentiation of several classes of neurons in vitro, including neural crest and placode-derived sensory neurons, dopaminergic neurons in the substantia nigra, basal forebrain cholinergic neurons, hippocampal neurons, and retinal ganglial cells. BDNF is expressed within peripheral ganglia and is not restricted to neuronal target fields, raising the possibility that BDNF has paracrine or even autocrine actions on neurons as well as non-neuronal cells.

ELISA: 1/500 - 1/2000. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: BDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR