Anti-Benzoylecgonine Mouse Monoclonal Antibody

Supplier: GeneTex
GTX15214
89360-110EA 673.73 CAD
89360-110
Anti-Benzoylecgonine Mouse Monoclonal Antibody
Antibodies
Benzoylecgonine is the predominant cocaine metabolite. This compound is not biologically active but is used as an indicator of cocaine use.

Type: Primary
Antigen: Benzoylecgonine
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR