Anti-B2M Mouse Monoclonal Antibody [clone: B2M-01]

Supplier: Genetex

GTX78315
89366-592EA 599.63 CAD
89366-592
Anti-B2M Mouse Monoclonal Antibody [clone: B2M-01]
Antibodies
Monoclonal Antibody to beta2-Microglobulin (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:200. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR