Anti-BAM1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX40638
89363-266EA 1152.52 CAD
89363-266
Anti-BAM1 Rabbit Polyclonal Antibody
Antibodies
Beta Amylase is an enzyme found in the seeds of higher plants and in sweet potatoes. It hydrolyses 1,4 alpha D glucosidic linkages in polysaccharides to remove successive maltose units from the non reducing ends of the chains. Used in the structural analysis of starch and glycogen.

Type: Primary
Antigen: BAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR