Anti-BMP8A Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX108555
89355-582EA 623.96 CAD
89355-582
Anti-BMP8A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to BMP8A (bone morphogenetic protein 8a)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: BMP8A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR