Anti-Bovine whole serum Sheep Polyclonal Antibody

Supplier: GeneTex
GTX77158
89366-184EA 286.36 CAD
89366-184
Anti-Bovine whole serum Sheep Polyclonal Antibody
Antibodies
This antiserum forms multiple arcs against normal bovine serum by immunoelectrophoresis and has strong anti-albumin.. Sheep were immunized with pooled normal bovine serum in Freund's adjuvant.

Type: Secondary
Antigen: Bovine Whole Serum
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR